Would you like to log in again?
Human Recombinant Chemerin (from E. coli)
Supplier: BioVision
AB124314-25UG
10006-006EA 467.33 USD InStock
10006-006
Human Recombinant Chemerin (from E. coli)
Proteins and Peptides
A chemoattractant and a ligand for G-protein coupled receptors
Recombinant human Chemerin has full biological activity when compared standards. The ED₅₀, as determined by its ability to chemoattract human Chem23R transfected BaF3 mouse pro-B cells, is typically 4-20 ng/mL.
Chemerin is a chemoattractant expressed in white adipose, liver and lung tissues. Chemerin is a ligand for the G-protein coupled receptor known as ChemR23 (or chemokine-like receptor-1), which is expressed mainly on dendritic cells, macrophages and some adipocytes. Recombinant human Chemerin, produced in E.coli, is a non-glycosylated protein containing 138 amino acids and having a total molecular mass of 16 kDa.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty