Would you like to log in again?
Human Recombinant Endostatin (from P. pastoris)
Supplier: BioVision
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
AB52022-100
10007-164EA 249.2 USD InStock
10007-164
Human Recombinant Endostatin (from P. pastoris)
Proteins and Peptides
>98% Pure active Recombinant human Endostatin.
BioVision’s human Endostatin is biologically active when compared to standards. The activity as calculated by ECE migration inhibition was found to be 50,000 IU/mg.
Endostatin has been identified as a C-terminal fragment of Collagen type 18, a recently identified member of a family of collagen-like proteins referred to as multiplexin family Endostatin specifically inhibits proliferation of endothelial cells although it does not affect the proliferation of EOMA cells. Endostatin also potently inhibits angiogenesis and tumor growth. Endostatin has an important role in endothelial cell adhesion and cytoskeletal organization. Endostatin can be found in vessel walls (elastic fibers) and basement membranes. Recombinant Endosatin expressed in yeast causes G1 arrest of endothelial cells, and endostatin treatment results in apoptosis of HUVE and HMVE cells.
DESCRIPTION:
Recombinant Human Endostatin produced in Pichia Pastoris is a single, glycosylated, polypeptide having a total molecular mass of 20,000 Dalton C-terminal fragment of collagen XVIII that has been shown to act as a potent inhibitor of angiogenesis and tumor growth in vitro and in vivo. Induces tyrosine phosphorylation of Shc (SH2 domain adapter protein) leading to specific inhibition in endothelial cell proliferation.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty