Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-SYN Rabbit Polyclonal Antibody

Supplier: BioVision
AB18814-100
10007-978EA 895 USD InStock
10007-978
Anti-SYN Rabbit Polyclonal Antibody
Antibodies
Synapsins are abundant brain proteins essential for regulating neurotransmitter release. All synapsins contain a short N-terminal domain that is highly conserved and phosphorylated by PKA or CaM kinase I. Phosphorylation of synapsin N-terminal domain at Ser9 inhibits its binding to phospholipids and diassociates synapsins from synaptic vesicles.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml

Type: Primary
Antigen: SYN
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty