Would you like to log in again?
K48
Supplier: BioVision
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
AB166765-50
10008-026EA 334.46 USD InStock
10008-026
K48
Proteins and Peptides
>95% Pure K48-linked Tri-Ubiquitin
Poly-ubiquitylation of target proteins through linkage at K48, is now the most thoroughly studied of the various chain linkages, and was once considered the hallmark of this post-translational modification. It is now clear that many, if not all, poly-Ub chain topologies likely play distinct and important roles in regulating cellular processes. Nevertheless, K48 linkage remains a critical pathway for the cells to maintain homeostasis through proteolytic degradation, and as such remains very intriguing for the study of DUBs that play a role in the degradation, as well as the proteasome itself. These tri-ubiquitin chains are generated from the enzymatic linkage (E2-25K) of wild-type ubiquitin through lysine 48. The most distal ubiquitin contains an arginine substitution for a lysine at position 48, limiting the chain length.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty