Would you like to log in again?
Human Recombinant Cyclophilin B (from E. coli)
Supplier: BioVision
AB285720-100UG
10008-158EA 750.79 USD InStock
10008-158
Human Recombinant Cyclophilin B (from E. coli)
Proteins and Peptides
>95% Pure Recombinant Active Human Cyclophilin B
Specific activity is > 220 nmoles/min/mg
Cyclophilin B is a cyclosporine-binding protein. It is both secreted and retained in the ER. When secreted, Cyclophilin B mediates chemotaxis and T cell adhesion to fibronectin. This is likely due to its prolyl cis/trans isomerase activity. Intracellularly, Cyclophilin B appears to serve as a molecular chaperone for molecules destined for secretion. It does so via stabilization, and facilitating the activity of additional chaperones. It also bind to cells derived from T- and B-lymphocytes, and may regulate cyclosporine A-mediated immunosuppression.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty