Would you like to log in again?
Giant keyhole limpet Native Hemocyanin-Keyhole Limpet (KLH)
Supplier: BioVision
AB285712-25MG
10008-330EA 357 USD InStock
10008-330
Giant keyhole limpet Native Hemocyanin-Keyhole Limpet (KLH)
Proteins and Peptides
Hemocyanins are proteins that use copper binding sites to bind and transport oxygen in a variety of arthropods and mollusks. Hemocyanin is isolated from the hemolymph of the animals. Hemocyanin is one of the strongest antigens known.
Hemocyanin can be use as an immunological reagent for many years. It is used as a carrier protein for antibody production against antigens. Recent advances in immunology and the role immune system plays in diseases have opened a whole new era of product development activities aimed at developing novel therapeutics for teaching the body's immune system to fight diseases. The approach involves the use of highly immunogenic molecule like the hemocyanin for non-specific immunostimulation (NSI) or active specific immunostimulation (ASI) using conjugate vaccines, wherein the tumor (disease) specific antigens are covalently bound to carrier protein like KLH and the product used in human clinical studies.
Native KLH has low endotoxin content and consistent high concentration in a phosphate buffer containing no extraneous metal ions for product stabilization, however with improved stability. This research grade material is suited for use in vaccine product development activities and also for routine immunological studies, antibody production, production of activated KLH and other developmental activities.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty