Would you like to log in again?
Human Recombinant BLMH (from E. coli)
Supplier: BioVision
AB168022-100UG
10008-506EA 781.41 USD InStock
10008-506
Human Recombinant BLMH (from E. coli)
Proteins and Peptides
>90% Pure Active Recombinant Human BLMH
Specific activity: > 1,000 pmole/min/µg. Measured by the hydrolysis of Met-AMC at pH 7.5, at 37°C.
BLMH is affiliate to the papain superfamily of the cysteine protease and the peptidase C1 family. BLMH is a cytoplasmic cysteine peptidase usually found as a homohexamer. The standard physiological role of BLMH has not been determined, but it shields normal and malignant cells from the glycopeptide antitumor drug BLM. BLMH catalyzes the inactivation of the antitumor drug BLM (a glycopeptide) by hydrolyzing the carboxyamide bond of its B-aminoalaninamide moiety and in addition demonstrates general aminopeptidase activity.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty