Would you like to log in again?
Recombinant Oxalate Decarboxy Active B (from E. coli)
Supplier: BioVision
AB285800-20UG AB285800-100UG AB285800-1MG
10009-670EA 467.29 USD InStock
10009-670 10009-668 10009-666
Recombinant Oxalate Decarboxy Active B (from E. coli)
Proteins and Peptides
>98% Pure bacterial Recombinant oxalate decarboxylase
Oxalate decarboxylase (OxdC, EC4.1.1.2) is a manganese-containing enzyme, which decomposes oxalic acid and oxalate. With OxdC catalysis, oxalate is split into formate and CO2. This enzyme belongs to the family of lyases, specifically the carboxy-lyases, which cleave carbon-carbon bonds. The systematic name of this enzyme class is oxalate carboxy-lyase (formate-forming). This enzyme is also called oxalate carboxy-lyase. The enzyme is composed of two cupin domains, each of which contains a Mn (II) ion. This enzyme participates in glyoxylate and dicarboxylate metabolism. This enzyme has been recognized for diagnostics in diverse biotechnological applications such as the clinical assay of oxalate in blood and urine, therapeutics, process industry, and agriculture to lower oxalate levels in foods and the environment. The recombinant protein made from the Bacillus Subtilis sequence includes OxdC with N-terminal His-tag.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty