Would you like to log in again?
CellExp™ Human Recombinant PCSK9 (from HEK293 Cells)
Supplier: BioVision
CellExp™
AB285803-20UG
10009-678EA 850.35 USD InStock
10009-678
CellExp™ Human Recombinant PCSK9 (from HEK293 Cells)
Proteins and Peptides
>97% Pure active Human Recombinant PCSK9
Measured by its ability to bind with human LDLR in a functional ELISA Immobilized recombinant human PCSK9 at 10 μg/ml (100 μl/well) can bind biotinylated human LDLR. The ED50 for this effect is typically 15 ng/ml.
Proprotein convertase subtilisin/kexin type 9 (PCSK9), is an enzyme which in humans is encoded by the PCSK9 gene. This gene encodes a proprotein convertase belonging to the proteinase K subfamily of the secretory subtilase family. This protein plays a major regulatory role in cholesterol homeostasis. PCSK9 binds to the epidermal growth factor-like repeat A (EGF-A) domain of the low-density lipoprotein receptor (LDLR), inducing LDLR degradation. PCSK9 may also have a role in the differentiation of cortical neurons. Mutations in this gene have been associated with a rare form of autosomal dominant familial hypercholesterolemia (HCHOLA3).
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty