Would you like to log in again?
Human Proteinase 3 Pr3 (from Neutrophils)
Supplier: BioVision
AB285815-25UG
10009-728EA 505.62 USD InStock
10009-728
Human Proteinase 3 Pr3 (from Neutrophils)
Enzymes
≥ 95% pure native Proteinase 3 Protein isolated from human neutrophils
Proteinase 3 is one of the serine proteinases localized in azurophil granules of polymorphonuclear leukocytes. It has been identified as the autoantigen of Wegener granulomatosis, a systemic vasculitis complicated by melomonocyte proliferation. It exhibits proteolytic activity towards a range of substrates. It is physiologically inhibited by alpha1-antitrypsin. The protein has been described as having antibiotic and growth-promoting properties and causing emphysema when administered by tracheal insufflation.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty