Would you like to log in again?
Human Native alpha-2-Macroglobulin (from Plasma)
Supplier: BioVision
AB285822-1MG
10009-744EA 497.21 USD InStock
10009-744
Human Native alpha-2-Macroglobulin (from Plasma)
Proteins and Peptides
≥ 95% pure native Alpha 2 Macroglobulin Protein isolated from human plasma
Alpha 2 Macroglobulin (A2M) is a plasma protease inhibitor which has been shown to exist in two forms. The Slow Form of A2M (S-A2M) is the form which possesses the ability to bind and inhibit proteases by a “trap” method. The Fast Form of A2M (F-A2M) is generated when S-A2M undergoes a conformational change due to either entrapment of a protease in the A2M bait region, or chemical cleavage of an internal thiol ester bond located near the bait region. F-A2M does not possess the ability to bind and inhibit protease activity. F-A2M is rapidly taken up by the liver, with a half-life of 2-4 minutes. In vivo, F-A2M typically represents only 0.17–0.7% of the total A2M in blood plasma of adults. The F-A2M plasma concentration is, however, increased in many disease states including pancreatitis, multiple sclerosis and sepsis. F-A2M has also been implicated in the inhibition of amyloid formation associated with Alzheimer’s disease and spongiform encephalopathy.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty