Would you like to log in again?
Human Recombinant WNT2 (from E. coli)
Supplier: BioVision
AB285937-1MG AB285937-50UG AB285937-10UG
10009-890EA 873.34 USD InStock
10009-890 10009-892 10009-894
Human Recombinant WNT2 (from E. coli)
Proteins and Peptides
>90% Pure Human Recombinant Wnt-2
This protein belongs to the Wnt family of proteins. The WNT gene family consists of structurally related genes that encode secreted signaling proteins involved in the Wnt signaling pathway. Wnt gene family members, including Wnt-1 through Wnt-10, play a key role in regulating cellular growth and differentiation. These proteins have been implicated in oncogenesis and in several developmental processes, including regulation of cell fate and patterning during embryogenesis. Wnt-2 is expressed in brain in the thalamus, in fetal and adult lung and in the placenta.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty