Would you like to log in again?
Human Recombinant CETP (active) (from Insect Cells)
Supplier: BioVision
AB285942-1MG AB285942-100UG AB285942-20UG
10009-918EA 551.62 USD InStock
10009-918 10009-914 10009-916
Human Recombinant CETP (active) (from Insect Cells)
Proteins and Peptides
>90% Pure Recombinant Human CETP.
The CETP Activity Assay uses a donor molecule containing a fluorescent self-quenched neutral lipid that is transferred to an acceptor molecule in the presence of CETP. CETP-mediated transfer of the fluorescent neutral lipid to the acceptor molecule results in an increase in fluorescence (Ex/Em: 465/535 nm).
Cholesteryl ester transfer protein (CETP), also called plasma lipid transfer protein, is a plasma protein that facilitates the transport of cholesteryl esters and triglycerides between the lipoproteins. It collects triglycerides from very-low-density (VLDL) or low-density lipoproteins (LDL) and exchanges them for cholesteryl esters from high-density lipoproteins (HDL), and vice versa. Inhibition of CETP has been proposed as a strategy to raise HDL cholesterol levels, a potential an approach to retarding atherogenesis and a strategy to reduce cardiovascular risk.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty