Would you like to log in again?
DuPont™ Tyvek® 400 Frocks with Laydown Collar and Pockets
Supplier: DuPont
Tyvek®
TY212SWHMD0030NF TY212SWHMD0008G1 TY212SWHXL003000 TY212SWHSM003000 TY212SWHLG0030VP TY212SWHSM003000 TY212SWHXL003000 TY212SWH5X0030NF TY212SWH2X003000 TY212SWH3X0008G1 TY212SWHLG003000 TY212SWH2X0030VP TY212SWH4X0008G1 TY212SWHXL0008G1 TY212SWH3X003000 TY212SWH4X0030NF TY212SWHMD003000 TY212SWH4X003000 TY212SWH7X003000 TY212SWHMD0030VP TY212SWHXL0030NF TY212SWH3X0030VP TY212SWH4X0030VP TY212SWH2X003000 TY212SWHXL0030VP TY212SWHLG003000 TY212SWH3X003000 TY212SWH6X003000 TY212SWH2X0008G1 TY212SWHMD003000 TY212SWHLG0008G1 TY212SWHSM0030NF TY212SWH3X0030NF TY212SWH5X003000 TY212SWH2X0030NF TY212SWH4X003000 TY212SWHLG0030NF
76188-318CS 54.33 USD InStock
CA10010-088 76188-318 76188-316 76188-118 CA10010-084 76188-116 76188-314 76188-336 76188-334 CA10010-086 76188-114 76188-112 76188-332 10016-932 76188-110 76188-330 10010-090 CA10010-082 10010-094 10010-092 10010-088 76188-328 10016-928 76188-108 76188-326 10010-086 76188-106 76188-324 CA10010-094 76188-322 76188-320 76188-120 10016-930 CA10010-090 CA10010-092 10010-084 10010-082
DuPont™ Tyvek® 400 Frocks with Laydown Collar and Pockets
Disposable Garments
Tyvek® 400 garments are composed of flash spun-high density polyethylene which creates a unique, nonwoven material available only from DuPont.
  • Laydown collar
  • Open wrists
  • Extends below hips
  • Front snap closure
  • Two front pockets
  • Serged seams
Tyvek® 400 provides an ideal balance of protection, durability and comfort of any limited use fabric technology. Tyvek® 400 fabric offers an inherent barrier against particles (down to 1.0 micron in size). Protection is built into the fabric itself; there are no films or laminates to abrade or wear away. Tyvek® 400 fabric's durability advantage over microporous film fabrics delivers consistently better barrier, even after wear and abrasion. Applications include lead and asbestos abatement/remediation, general maintenance/operations, spray painting, general clean-up.
Lab coats feature body-length jacket design with front snap closures for easy donning and doffing. Ideal for applications that do not require the full body coverage of a coverall. Lab coats also feature a laydown collar and two front pockets for holding writing instruments, small tools or parts.
Certifications: Thickness meets ASTM D1777.
Order Now
ORDER
SPECIFICATIONS
COMPLEMENTARY PRODUCTS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty