Would you like to log in again?
DuPont™ Tychem® 4000 Coveralls with Standard Hood, Bound Seams
Supplier: DuPont
Tychem®
SL127BWH2X001200 SL127BWH4X001200 SL127BWHMD001200 SL127BWHXL001200 SL127BWH2X001200 SL127BWHLG001200 SL127BWHMD001200 SL127BWH7X001200 SL127BWHXL001200 SL127BWH6X001200 SL127BWH3X001200 SL127BWHLG001200 SL127BWH5X001200 SL127BWH4X001200 SL127BWH3X001200
10010-176CS 474.64 USD InStock
10010-176 CA10010-176 10010-174 CA10010-178 10010-168 CA10010-172 10010-178 CA10010-174 CA10010-168 10016-494 CA10010-170 10010-172 10016-490 10016-492 10010-170
DuPont™ Tychem® 4000 Coveralls with Standard Hood, Bound Seams
Disposable Garments
DuPont™ Tychem® 4000 delivers effective protection against a range of chemical environments.
  • Standard fit hood
  • Elastic wrists and ankles
  • Storm flap with adhesive closure
  • Bound seams
Tychem® 4000, utilizes Saranex™ 23-P film laminated to Tyvek® fabric to provide a rugged and durable fabric that resists abrasion. Tychem® 4000 fabric provides at least 30 minutes of protection against >124 chemical challenges. Typical applications: chemical mixing, hazardous remediation, emergency medical response, paint spraying, and nuclear environments. Tightly sewn seam is covered with garment fabric to reinforce seam and reduce potential for particle penetration. Coveralls feature an attached hood with elastic around face opening and elastic openings at the wrists and ankles for a tighter fit. Storm flap covers zipper, which can be sealed by the wearer with adhesive strip to prevent intrusion at zipper.
Certifications: Thickness meets ASTM D1777.
Order Now
ORDER
SPECIFICATIONS
COMPLEMENTARY PRODUCTS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty