Would you like to log in again?
DuPont™ Tychem® 2000 Coveralls with Standard Hood and Attached Socks, Bound Seams
Supplier: DuPont
Tychem®
QC122BYL2X0012BN QC122BYL4X001200 QC122BYLXL0012BN QC122BYL3X001200 QC122BYLLG001200 QC122BYL5X001200 QC122BYLMD001200 QC122BYL2X001200 QC122BYL6X001200 QC122BYLLG0012BN QC122BYLXL001200 QC122BYL3X001200 QC122BYLMD001200 QC122BYLXL001200 QC122BYLLG001200 QC122BYL6X0012BN QC122BYL5X0012BN QC122BYL4X001200 QC122BYLMD0012BN QC122BYL3X0012BN QC122BYL2X001200 QC122BYL4X0012BN
10010-242CS 293.71 USD InStock
10010-242 CA10010-242 10010-240 CA10010-244 10010-246 10010-244 CA10010-240 CA10010-238 10010-238 10010-248 CA10010-246 CA10010-248 89207-848 10016-384 10016-374 10016-386 10016-376 10016-378 10016-380 10016-370 10016-382 10016-372
DuPont™ Tychem® 2000 Coveralls with Standard Hood and Attached Socks, Bound Seams
Disposable Garments
A lightweight, and durable fabric, DuPont™ Tychem® 2000 utilizes the strength of DuPont™ Tyvek® fabric and a polyethylene coating.
  • Standard fit hood
  • Elastic wrists
  • Attached socks
  • Storm flap with adhesive closure
  • Bound seams
Tychem® 2000 fabric provides at least 30 minutes of protection against 42 chemical challenges. Tychem® 2000 is used for light splash protection in a variety of industrial environments, including petroleum refining, pulp and paper manufacturing, food processing, chemical processing, and pharmaceutical manufacturing. Tightly sewn seams are covered with garment fabric to reinforce seam and reduce potential for particle penetration. Attached hood with elastic around face opening is standard fit. Storm flap covers zipper, which can be sealed by the wearer with adhesive strip to prevent intrusion at zipper. Elastic openings at wrists allow for a tighter fit. Integrated socks are composed of the same garment material.
Certifications: Thickness meets ASTM D1777.
Order Now
ORDER
SPECIFICATIONS
COMPLEMENTARY PRODUCTS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty