Would you like to log in again?
Orion™ Standard Epoxy DO Probe for Lab or Field, Thermo Scientific
Supplier: Thermo Scientific
Orion™
081010MD 081010MD
10010-708EA 1028.14 USD InStock
CA10010-708 10010-708
Orion™ Standard Epoxy DO Probe for Lab or Field, Thermo Scientific
Electrodes Dissolved Oxygen Electrodes
Use this product as the name implies, either in the laboratory or field.
  • Designed for use with star series meters
  • Less than 1% drift per day
  • 90% of final value in 10 seconds
  • Minimum sample flow of 20 cm/second
The rugged body and versatility of the Orion™ Standard Epoxy DO Probe ensures reliable results regardless of the environment.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty