Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.

Orion™ DO Probe for Lab or Field, Thermo Scientific

Supplier: Thermo Scientific
Orion™
083005MD 083010MD 083060MD 083005MD 083025MD
10010-710EA 1574.8 USD InStock
CA10010-712 10010-710 10010-716 10010-714 CA10010-710
Orion™ DO Probe for Lab or Field, Thermo Scientific
Electrodes Dissolved Oxygen Electrodes
Use this dissolved oxygen probe as the name implies, either in the laboratory or filed.

  • Less than 1% drift per day
  • Maximum overpressure is 6 bar
  • Minimum sample flow of 10 cm/second
  • Oxygen consumption is 0.008 µg/h (mg/L) at 20°C
  • 90% of final value in 10 seconds
  • Dual, thermally separated sensors

The Orion™ DO Probe for Laboratory or Field is durable and accurate, featuring automatic temperature compensation and convenient screw-on membrane caps.

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty