Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.

Orion™ AUTO-STIR™ BOD Probe, Thermo Scientific

Supplier: Thermo Scientific
Orion™
086030MD 086030MD
10010-718EA 2354.16 USD InStock
10010-718 CA10010-718
Orion™ AUTO-STIR™ BOD Probe, Thermo Scientific
Electrodes Dissolved Oxygen Electrodes
Accurately and conveniently conduct dissolved oxygen measurements with the Orion™ AUTO-STIR™ BOD probe.

  • Designed for use with Orion Star and Star Plus Serie Benchtop Models
  • Less than 1% drift per day
  • Maximum overpressure is 6 bar
  • Minimum sample flow of 10 cm/second
  • Oxygen consumption is 0.008 µg/h (mg/L) at 20°C
  • 90% of final value in 10 seconds
  • Dual, thermally separated sensors

Self-stirring with One-Touch control, this highly efficient, integrated stirrer eliminates oxygen stratification in BOD bottles.

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty