Would you like to log in again?
Orion™ Epoxy ATC Probe, Thermo Scientific
Supplier: Thermo Scientific
Orion™
927005MD 917005 927005
34104-398EA 684.75 USD InStock
34104-398 10010-740 34104-260
Orion™ Epoxy ATC Probe, Thermo Scientific
Electrodes Other Electrodes
Get fast, accurate measurements in the laboratory or field.
The Orion™ Epoxy ATC Probe automatically corrects pH measurements for variation in electrode slope due to temperature change.
Ordering information: For temperature readings only. pH probe is sold separately.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty