Would you like to log in again?
Orion™ Glass ATC Probe, Thermo Scientific
Supplier: Thermo Scientific
Orion™
927006 917006 927006MD
34104-255EA 666.59 USD InStock
34104-255 10010-742 34104-262
Orion™ Glass ATC Probe, Thermo Scientific
Electrodes pH Electrodes
Automatically correct pH measurements for variation in electrode slope due to temperature change with the Orion™ Glass ATC Probe
  • pH range of 0 to 14
Three different models are available to accommodate different Orion™ meters. The Mini DIN version is compatible with older meters with use of an adapter cable.
Available with three different connector options.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty