Would you like to log in again?
Orion™ AquaPro™ pH Combination Electrodes, Thermo Scientific
Supplier: Thermo Scientific
9107APMD 9135APWP 9156APWP 9104APWP
10010-792EA 656.78 USD InStock
10010-792 10010-790 10010-782 14004-266
Orion™ AquaPro™ pH Combination Electrodes, Thermo Scientific
Electrodes pH Electrodes
Read viscous liquid samples quickly and accurately without the sample coming into contact with silver ions with Orion™ AquaPro™ pH Combination Electrodes.
  • Always store in Orion™ pH Electrode Storage Solution
  • Either store in a beaker of storage solution or reassemble in the provided electrode storage bottle
  • pH range is 0 to 14
Orion™ AquaPro™ pH Combination Electrodes have a clog-resistant and open double junction reference system. These electrodes are ideal for measurements in samples containing proteins or TRIS buffers, which do not contain mercury. The electrodes require minimal maintenance because there is no fill solution; instead they have a polymer gel reference.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty