Would you like to log in again?
Orion™ Chloride Electrodes, Thermo Scientific
Supplier: Thermo Scientific
Orion™
9417BN 9617BNWP 9617BNWP
34116-764EA 1275.16 USD InStock
34116-764 CA10010-812 10010-812
Orion™ Chloride Electrodes, Thermo Scientific
Electrodes ISE Electrodes
Measure free chloride in the range of 1 x 10−5M to 1M with Orion™ Chloride Electrodes.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty