Would you like to log in again?
Orion™ Bromide Electrodes, Thermo Scientific
Supplier: Thermo Scientific
Orion™
9635BNWP
10010-816EA 1489.93 USD InStock
10010-816
Orion™ Bromide Electrodes, Thermo Scientific
Electrodes ISE Electrodes
These electrodes are designed for measuring bromide ions in aqueous solutions quickly, simply, and economically.
  • Stable, drift-free potentials
  • Sure-Flow® junction allows easy cleaning and is easy to use in tough samples
  • When conventional electrode junctions clog, the Sure-Flow® junction can be opened and flushed clean simply by pressing the cap
  • Half-cell electrode used in conjunction with the double junction reference electrode offers the benefits of the Sure-Flow® junction design
Ordering information: The half cell electrodes require a separate reference electrode which can be ordered separately. Screw cap electrode is supplied without cable.
Order Now
ORDER
ACCESSORIES
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty