Would you like to log in again?
Human Recombinant Sirt6 His Tagged Active (from E. coli)
Supplier: BioVision
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
AB285957-1MG AB285957-100UG AB285957-20UG
10011-096EA 398.36 USD InStock
10011-096 10011-092 10011-094
Human Recombinant Sirt6 His Tagged Active (from E. coli)
Proteins and Peptides
≥90% Pure, Active, Recombinant, AOF, His tagged human SIRT6 (NAD-dependent Deacetylase Sirtuin-6, Mono-ADP-ribosyltransferase Sirtuin 6, SIR2-like Protein 6, SIR2L6).
  • >3 mU/mg
Human Sirtuins function as intracellular regulatory proteins with mono-ADP-ribosyltransferase activity. Sirtuin 6 (SIRT6) is a NAD-dependent protein deacetylase with activity towards Lys-9 and Lys-56 of histone H3. It modulates acetylation of histone H3 in telomeric chromatin during the S-phase of the cell cycle. It is required for genomic stability and for normal IGF1 serum levels and normal glucose homeostasis. It also regulates cellular senescence and apoptosis. Unlike most sirtuins, SIRT6 preferentially removes long-chain fatty-acyl, rather than acetyl, protein modifications. This activity regulates secretion of the inflammation-associated protein TNF-α.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty