Would you like to log in again?
Axygen® Microcentrifuge Screw Caps, Corning
Supplier: Corning
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Axygen®
SCO-R SCO-BR SCO-G SCO-LP-BR SCO-C SCO-V SCO-LP-C SCO-B SCO-LP-B SCO-Y SCO-LP-Y
10011-472CS 970.7 USD InStock
10011-472 10011-460 10011-430 10011-440 10011-432 10011-442 10011-434 10011-466 10011-444 10011-468 10011-428
Axygen® Microcentrifuge Screw Caps, Corning
Closures Test/Sample Tube Closures
These caps are compatible with all Axygen Scientific screw top microcentrifuge tubes, including both skirted and nonskirted variations of the 0.5 ml, 1.5 ml, and 2 ml microcentrifuge tubes.
  • Available looped and non-looped
The O-ring closure is formulated and manufactured from a special compound resistant to chemicals such as anhydrous ethyl alcohol, purified ether, hexane, acetone, chloroform, acetonitrile, 40% trifluoroacetic acid solution, and other chemicals used in most laboratories. The caps have extra long lips to help protect samples and ribs to allow one hand operation.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty