Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 25, 2024. During this time, our site will be not available. We appreciate your patience.
Axygen® Screw Top Microcentrifuge Tubes without Caps, Polypropylene, Corning
Supplier: Corning
Axygen®
ST-150-X ST-200-SS ST-150-SS ST-200 ST-150-C-S ST-45-SS ST-200-SS-X ST-200-X ST-150 ST-050 ST-150-SS-X ST-050-SS ST-050-SS-X ST-200-C-S ST-050-X
47751-114CS 708.06 USD InStock
47751-114 47751-112 89179-786 10011-670 10011-662 10011-674 10011-652 10011-660 10011-672 10011-666 10011-656 10011-664 10011-654 10011-668 10011-658
Axygen® Screw Top Microcentrifuge Tubes without Caps, Polypropylene, Corning
Tubes Microcentrifuge Tubes
Polypropylene screw cap microcentrifuge tubes feature an alignment tab to align the tube with the centrifuge rotor for multiple applications, such as desalting.
This feature allows the tube to be re-spun without disturbing the partially pelletized sediment.
Certifications: Certified RNase-, DNase-, and pyrogen-free.
Ordering information: For compatible screw caps, see the 10011-394 series.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty