Would you like to log in again?
Axygen® Research-Grade Pipette Tips, Corning
Supplier: Corning
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Axygen®
T-1000-C-R-S T-1005-WB-C T-1000-C T-1000-C-R
10011-916CS 161.92 USD InStock
10011-916 10011-310 10011-306 10011-308
Axygen® Research-Grade Pipette Tips, Corning
Pipette Tips
These tips are designed to fit a wide variety of 1000 µl single-channel pipettors.
Flexible walls and a series of internal sealing rings ensure a secure fit and less force required to load and eject. Accuracy and precision testing is performed for every manufacturing lot using the finest pipettors in the industry. Wide bore tips are ideal for pipetting viscous materials, genomic DNA or delicate cell cultures.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty