Would you like to log in again?
Axygen® Research-Grade Pipette Tips, Corning
Supplier: Corning
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Axygen®
T-400-R MTX-1250-C-R T-350-C T-200-Y-R-S T-200-Y T-300 T-350-C-R TXL-10-R T-400 T-205-WB-C T-200-C-R T-300-STK T-205-WB-C-R-S T-300-R T-1000-B-R T-1000-B T-200-Y-R TR-222-C-R-S T-300-STK-S TXL-10 MTX-1250-C T-200-C T-1005-WB-C-R T-1005-WB-C-R-S TR-222-C T-300-R-S TR-222-C-R T-200-C-R-S TXL-10-R-S T-205-WB-C-R T-1000-B-R-S
47745-486CS 694.02 USD InStock
47745-486 47745-484 10011-918 47745-488 10011-340 10011-320 10011-344 10011-322 10011-300 47734-724 10011-302 10011-324 10011-920 10011-326 10011-304 10011-922 10011-928 10011-372 10011-374 12000-742 12000-744 10011-930 10011-876 10011-312 10011-874 10011-336 10011-314 10011-316 10011-338 10011-932 10011-318
Axygen® Research-Grade Pipette Tips, Corning
Pipette Tips
Tips are designed to fit a wide variety of single-channel and multichannel pipettors.
  • RNase-, DNase-, and endotoxin-free
Presterilized tips are exposed to e-beam radiation for protection from contamination.
Flexible walls and a series of internal sealing rings ensure a secure fit and less force required to load and eject. Accuracy and precision testing is performed for every manufacturing lot using the finest pipettors in the industry.
Ordering information: Graduated tips have graduations at 10, 50, and 100 µl. The 200 µl wide-bore tips have a 1.52 mm (¹/₁₆") orifice, and 1000 µl wide-bore tips have a 2.0 mm ¹³/₁₆"
orifice. Both options for 30 µl tips and the 60 µl tips are designed for use with 16-channel pipettors for either 384- or 1536-well pipetting.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty