Would you like to log in again?
Axygen® Research-Grade Bevelled Reference Pipette Tips 200 µl, Corning
Supplier: Corning
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Axygen®
TR-222-Y TR-222-Y-R TR-222-C-STK TR-222-Y-R-S TR-222CLSTKS TR-222-Y-STK TR-222-C-STK-S
95042-896CS 578.66 USD InStock
95042-896 10011-894 10011-376 47734-722 10011-378 47734-742 10011-934
Axygen® Research-Grade Bevelled Reference Pipette Tips 200 µl, Corning
Pipette Tips
Axygen tips are designed to fit a wide variety of single-channel and multichannel pipettors.
Flexible walls and a series of internal sealing rings ensure a secure fit and less force required to load and eject. Accuracy and precision testing is performed for every manufacturing lot using the finest pipettors in the industry.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty