Would you like to log in again?
Human Recombinant AKT1 active (from Baculovirus (Sf9 Insect cells))
Supplier: BioVision
AB62279-5
10011-950EA 386.92 USD InStock
10011-950
Human Recombinant AKT1 active (from Baculovirus (Sf9 Insect cells))
Proteins and Peptides
>90% Pure active Recombinant Akt1.
Akt1 or Protein Kinase B α(PKBα) is a serine/threonine kinase that belongs to the Akt family. Akt1 is activated in cells in response to diverse stimuli such as hormones, growth factors and extracellular matrix components and is involved in glucose metabolism, transcription, survival, cell proliferation, angiogenesis, and cell motility (1). The PI3K generates phosphatidylinositol-3,4,5-trisphosphate (PIP3), a lipid second messenger essential for the translocation of Akt1 to the plasma membrane where it is phosphorylated and activated by phosphoinositide-dependent kinase-1 (PDK-1) (2) and phosphoinositide-dependent kinase-2 (possibly ILK) (3). Akt1 has numerous cellular substrates including proteins, which promote the inhibition of apoptosis such as the Forkhead transcription factors and the Bcl-2 family member Bad (4). In addition, the cyclin dependent kinase inhibitors are substrates of Akt which when phosphorylated relinquish their inhibitory influence on cell cycle progression. Akt also mediates many of the stimulatory effects of insulin on glucose metabolism through deactivation of glycogen synthase kinase, activation of phosphofructo-kinase, and modulation of glucose transporter activity. Akt1 has been shown to play a role in human cancers (4). It is frequently overexpressed and active in many types of human cancers including cancers of colon, breast, brain, pancreas and prostate as well as lymphomas and leukemias (5). Akt1 plays an important role in cancer cell survival and proliferation thereby contributing to cancer progression.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty