Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Hemoccult® Guaiac Fecal Occult Blood Test Systems, Beckman Coulter®

Supplier: Beckman Coulter

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

Hemoccult®
60152 61100 61100 61130A 63202A 60151A 60151A 63202A 61200A 61130A
10012-000CS 1574.25 USD InStock
CA74020-016 10012-000 CA74020-018 10012-012 10012-002 10012-004 CA74020-051 80078-660 CA74020-054 CA74020-012
Hemoccult® Guaiac Fecal Occult Blood Test Systems, Beckman Coulter®
Clinical Diagnostic Supplies Clinical Reagents and Kits
Qualitative procedure for detecting fecal occult blood uses a guaiac paper slide test.

  • Qualitative
  • Pre-qualified
  • Lot to lot consistency

The rigorous pre-qualification of raw materials and a unique quality control system allows the manufacturing of lot to lot consistency that is unmatched in the industry.

Certifications: CLIA waived.

Ordering information: Single Slides pack 10012-000 is supplied with 100 slides, 100 applicator sticks, and two 15mL bottles of developer. Single Slides 80078-660 is packaged as a case that includes 1000 slides, 1000 applicator sticks, and twenty 15mL bottles of developer. Dispensapak™ system is supplied with 100 patient kits (triple slide and applicator sticks only) and six 15mL bottles of developer. Dispensapak™ Plus system is supplied with 40 patient kits and two 15mL bottles of developer. Patient Pak system includes 34 Hemoccult II® triple slides (test cards), two 15mL bottles of developer, and 102 applicator sticks. Hemoccult® Tape includes two tape dispensers and two 15mL bottles of developer.

Order Now
 

ORDER

ACCESSORIES

DOCUMENTATION

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty