Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 25, 2024. During this time, our site will be not available. We appreciate your patience.
Pipettor Holder, Acrylic, Angled, 3 Slot or 6 Slot, TrippNT
Supplier: TrippNT
50071 50176
10012-826EA 79.53 USD InStock
10012-826 10012-830
Pipettor Holder, Acrylic, Angled, 3 Slot or 6 Slot, TrippNT
Pipette Stands
Comfortable access to auto pipettors stored at an angle in these 3 or 6 position acrylic holders.
  • Stores auto pipettors at a comfortable angle for use and storage
  • Choose from 2 sizes
  • Acrylic construction
  • Silicone bumpers to prevent sliding on bench or countertop
  • In stock and ready to ship
  • American made by TrippNT
  • Total replacement Lifetime Guarantee
Super glossy white or blue acrylic these holders are designed for safe storage and a comfortable reach and return of automatic pipettors.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty