Would you like to log in again?
Hood or Wall Mount Pipette Storage Bins, PVC, TrippNT
Supplier: TrippNT
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
50428 50869
10012-832EA 468.13 USD InStock
10012-832 10012-834
Hood or Wall Mount Pipette Storage Bins, PVC, TrippNT
Pipette Stands
Wall mountable angled large pipette organizer with 5 compartments keeps expensive pipettes clean, safe and available.
  • Stores expensive volumetric pipettes safely
  • Angled bins secure and safely store pipettes
  • Provides convenient access to pipettes
  • Tape or magnet mount
  • White PVC or acrylic construction
  • American made by TrippNT
  • Total replacement Lifetime Guarantee
Installs instantly with tape or magnet. Angled compartments keep pipettes with tip down with large opening to make access easy. Completely rust proof PVC construction. Bright white color gives a clean look and feel to the area.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty