Would you like to log in again?
StemBoost™ 2i-Reprogramming Cocktail Set (1000X), Sterile-Filtered, BioVision
Supplier: BioVision
AB286238-5ML AB286238-1ML
10013-820EA 1080.18 USD InStock
10013-820 10013-096
StemBoost™ 2i-Reprogramming Cocktail Set (1000X), Sterile-Filtered, BioVision
Cell Culture Supplements and Additives
A convenient, sterile-filtered cocktail solution (1000X, in DMSO) containing two small molecules used for enhancing the reprogramming efficiency of human somatic cells to induced pluripotent stem cells (iPSCs).
Caution: For research use only. Not for use in humans.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty