Would you like to log in again?
Recombinant Turbonuclease (from E. coli)
Supplier: BioVision
AB286000-50000UNITS
10013-242EA 834.19 USD InStock
10013-242
Recombinant Turbonuclease (from E. coli)
Enzymes
Pure recombinant active bacterial TurboNuclease
BioVision’s TurboNuclease is a recombinant form of Serratia macescens extracellular endonuclease (encoded by the same gene of Benzonase) produced in E. coli using a proprietary process. This nonspecific endonuclease hydrolyzes both single- and double-stranded nucleic acids (DNA and RNA) to 5’-phosphorylated oligonucleotides of 1-4 bases in length. TurboNuclease is a highly purified homodimer of 27 kDa subunits that has exceptional high specific activity and is free of protease activity. It is ideal to digest nucleic acids and to reduce viscosity during protein purification and sample preparation.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty