Would you like to log in again?
Caspselect Caspase-3 Immunoassay Kit, BioVision
Supplier: BioVision
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
AB234045-100TEST
10013-626EA 1348.31 USD InStock
10013-626
Caspselect Caspase-3 Immunoassay Kit, BioVision
Assays Protein Assays
Specifically detects Caspase-3
  • Fluorescence (Ex/Em 370-425/ 490-525 nm)
  • Mammalian
  • 100 assays
  • Effective immunosorbent enzyme assay for specific, quantitative detection of caspase-3 activity in microtiter plates
Activation of caspase-3 plays a key role in initiation of cellular events during apoptosis. The CaspSELECT™ Caspase-3 Immunoassay Kit provides an effective immunosorbent enzyme assay for specific, quantitative detection of caspase-3 activity in microtiter plates. The assay utilizes caspase-3 polyclonal antibody to capture activated caspase-3 from cell lysates. Caspase substrate DEVD-AFC is then added and is cleaved proportionally to the amount of activated caspase-3 in the cell lysate. The cleavage generates free AFC which can be analyzed fluorometrically (Ex./Em. = 400 nm/505 nm) using a fluorescence plate reader. The assay ensures absolute specific detection of caspase-3. Other known caspases and non-specific proteases are not detected.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty