Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Beta-Secretase Activity Assay Kit, BioVision

Supplier: BioVision
AB65357-100
10013-686EA 1392 USD InStock
10013-686
Beta-Secretase Activity Assay Kit, BioVision
Assays Protein Assays
Highly Sensitive Assay

  • Detection method- Fluorescence (Ex/Em 335-355/ 495-510 nm)
  • Species reactivity- Mammalian
  • Applications- Convenient fluorescence method for detecting β-secretase activity in biological and purified samples.

β-Secretase has been implicated to be an excellent target for anti-amyloid therapy for the treatment of Alzheimer’s disease. The β-Secretase activity Assay Kit provides a convenient fluorescence method for detecting β-secretase activity in biological and purified samples. The assay utilizes a secretase-specific peptide conjugated to two reporter molecules EDANS and DABCYL. In the uncleaved form, the fluorescent emissions from EDANS are quenched by the physical proximity of the DABCYL moiety. Cleavage of the peptide by secretase physically separates EDANS and DABCYL allowing for the release of a fluorescent signal. The level of secretase enzymatic activity in samples is proportional to the level of fluorescence intensity.

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty