Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Hdac3 Inhibitor Screening Kit, BioVision

Supplier: BioVision
AB283379-100TEST
10013-688EA 1149.12 USD InStock
10013-688
Hdac3 Inhibitor Screening Kit, BioVision
Assays Protein Assays
Sensitive Assay, HTS

  • Detection method- Fluorescence (Em/Ex = 380/500 nm)
  • Applications- Suitable for high throughput screening of HDAC3 inhibitors

Histone deacetylases (HDACs) represent a large family of enzymes identified as key regulators of nucleosomal histone acetylation, a major event that controls eukaryotic gene transcription and are classified into three groups. They are believed to be involved in important biological activities, such as cell differentiation, proliferation, apoptosis, and senescence. In BioVision’s HDAC-3 Inhibitor screening Kit, HDAC-3 and developer will deacetylate and cleave the substrate [Arg-His-Lys-Lys(Ac)-AFC] to release the quenched fluorescent group (AFC), which can be detected at Em/Ex=380/500 nm. In presence of a HDAC-3 inhibitor, the cleavage will be inhibited. The kit provides a rapid, simple, sensitive, and reliable test, which is also suitable for high throughput screening of HDAC-3 inhibitors. Trichostatin A (TSA) is included as a control inhibitor to compare the efficacy of the test inhibitors.

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty