Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Urine Albumin-to-Creatinine Ratio Uacr as Kit, BioVision

Supplier: BioVision

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

AB241018-100TEST
10013-754EA 1254.45 USD InStock
10013-754
Urine Albumin-to-Creatinine Ratio Uacr as Kit, BioVision
Assays Protein Assays
Only kit on the market to measure UACR

  • Detection method- Fluorescence (Ex/Em = 600/630 nm) & Absorbance (OD 570 nm)
  • Applications- Estimation of albumin in biological samples
  • - Estimation of creatinine in biological samples
  • - Determination of UACR in mammalian urine samples

Urine Albumin-to-Creatinine Ratio (UACR) is one of the two markers used to determine chronic kidney disease (CKD). UACR is recommended to be measured on regular basis on people living with Type I and Type II diabetes. UACR is defined as the ratio between albumin (reported in mg/dl) and creatinine (reported in g/dl). This ratio estimates the amount of albumin excreted in urine during a 24 hr period. Albuminuria is diagnosed when UACR is greater than 30 mg albumin/g creatinine. BioVision’s UACR Assay Kit provides a simple, sensitive, and high-throughput adaptable assay that detects albumin (detection range: 0.02- 2.5 mg/ml), creatinine (detection range: 0.002 -0.5 mg/ml) and urine albumin-to-creatinine ratio. The UACR ratio is determined in two steps: First, albumin is determined by using a probe (AB580) that specifically recognizes albumin (Ex/Em = 600/630 nm). Second, creatinine is converted to sarcosine via enzymatic reactions. Sarcosine is specifically oxidized generating a product that reacts with a probe producing a chromophore that can be detected at 570 nm.

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty