Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

HMG-CoA Reductase Activity/Inhibitor Screening Kit (Colorimetric), BioVision

Supplier: BioVision

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

AB204701-100TEST
10013-762EA 1165.7 USD InStock
10013-762
HMG-CoA Reductase Activity/Inhibitor Screening Kit (Colorimetric), BioVision
Assays Protein Assays
Simple assay to measure activity of purified enzyme and screening inhibitors/activators of HMGR

  • Detection method- Absorbance (OD 340 nm)
  • Applications- Measurement of HMG-CoA reductase activity
  • - Screening/studying/characterizing HMG-CoA reductase inhibitors/activators

HMG-CoA reductase (3-hydroxy-3-methyl-glutaryl-CoA reductase or HMGR) (EC 1.1.1.34) is the rate-controlling enzyme of the mevalonate pathway, the metabolic pathway that produces cholesterol from acetyl-CoA. In an NADPH-dependent reaction, HMG-CoA reductase reduces HMG-CoA to generate mevalonate and CoA. The enzyme is target of a group of cholesterol-lowering drugs known as statins. Inhibition of HMG-CoA reductase induces expression of LDL receptors in the liver, which lowers plasma concentration of cholesterol. BioVision’s HMG-CoA Reductase Activity Assay Kit measures utilization of NADPH, which can be measured by the decrease of absorbance (OD 340 nm). The kit is suitable for measuring activity of purified enzyme and screening inhibitors/activators of HMG-CoA reductase. The limit of detection is below 0.05 mU.

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty