Would you like to log in again?
SEPTA.HS WHITE SIL/FOIL 1,000 CASE
Supplier: MicroSolv Technology Corp
Septa, white silicone/ foil, for crimp caps for headspace. 45 Shore, 3mm thick. 1, 000/case^
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
SEPTA.HS WHITE SIL/FOIL 1,000 CASE 10014-052 95025-14 Case of 1 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty