Would you like to log in again?
False Bottom Tubes and Caps, Globe Scientific
Supplier: Globe Scientific
5528B 5527 5527B
10014-268PK 107.72 USD InStock
10014-268 10014-262 10014-260
False Bottom Tubes and Caps, Globe Scientific
Tubes Test Tubes
False bottom tubes are designed for use as a secondary tube for barcoding on automated analyzers.
Tubes may be used with a variety of popular analyzers, including Hitachi®, Olympus®, Corning®, Ortho®, Kodak®, Beckman®, IA®, Tosoh®, Vitros®, and more.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty