Would you like to log in again?
GFP CERTIFIED® FLUOFORTE® Reagent (Non-AM), Enzo Life Sciences
Supplier: Enzo Life Sciences
ENZ-52555-0250
10014-480EA 387.39 USD InStock
10014-480
GFP CERTIFIED® FLUOFORTE® Reagent (Non-AM), Enzo Life Sciences
Protein Linking and Labeling Systems
Calcium dye.
The GFP CERTIFIED® FLUOFORTE®Reagent (Non-AM) is the most sensitive fluorescent calcium indicator relative to other commercially available red fluorescent dyes. It can be used for detecting intracellular calcium mobilization across a broad spectrum of biological targets. Esterase hydrolysis is not required for live cell analysis via flow cytometry or fluorescence microscopy.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty