Would you like to log in again?
AMPIGENE® DNA Ladder 50 to 1500 bp, Enzo Life Sciences
Supplier: Enzo Life Sciences
GEN102-0100
10014-496EA 127.42 USD InStock
10014-496
AMPIGENE® DNA Ladder 50 to 1500 bp, Enzo Life Sciences
Electrophoresis Reagents Molecular Weight Markers
Ready to use DNA standards for quantifying PCR yield
For size determination and DNA quantification using agarose electrophoresis
AMPIGENE® DNA Ladder 50-1,500 bp contains DNA fragments ranging from 50bp to 1,500bp that can be used as molecular weight standards for agarose gel electrophoresis. The bands are evenly spaced and are intensified at 200bp, 500bp and 1,200bp to serve as easily identifiable reference bands. The approximate mass of DNA in each of the bands is provided, assuming 5µl load, for determining the approximate mass of DNA in samples with comparable intensity and size.
Ordering information: Contains Ready-to-use AMPIGENE® DNA Ladder 50-1,500 bp and Sample Loading Buffer.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty