Would you like to log in again?
PBS (Endotoxin-Free), Enzo Life Sciences
Supplier: Enzo Life Sciences
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
505-007-LD15 505-007-L010
10014-562EA 153.81 USD InStock
10014-562 89151-648
PBS (Endotoxin-Free), Enzo Life Sciences
Buffers Cell Culture Buffers
Sterile and endotoxin-free PBS for use with TLRgrade™ or endotoxin-free grade reagents.
Product has been subjected to multiple rounds of LPS removal by adsorption with activated charcoal.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty