Would you like to log in again?
AMPIGENE® DNA Ladder 100 to 10000 bp, Enzo Life Sciences
Supplier: Enzo Life Sciences
GEN104-0100
10014-580EA 136.6 USD InStock
10014-580
AMPIGENE® DNA Ladder 100 to 10000 bp, Enzo Life Sciences
Electrophoresis Reagents Molecular Weight Markers
Ready to use DNA standards for quantifying PCR yield
For size determination and DNA quantification using agarose electrophoresis
AMPIGENE® DNA Ladder 100-10,000 bp contains DNA fragments ranging from 100bp to 10,000bp that can be used as molecular weight standards for agarose gel electrophoresis. The bands are evenly spaced and are intensified at 500bp, 1,500bp and 3,000 to serve as easily identifiable reference bands. The approximate mass of DNA in each of the bands is provided, assuming 5µl load, for determining the approximate mass of DNA in samples with comparable intensity and size.
Ordering information: Contains Ready-to-use AMPIGENE® DNA Ladder 100-10,000 bp and Sample Loading Buffer.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty