Would you like to log in again?
Adipogenesis Assay Kit, Enzo Life Sciences
Supplier: Enzo Life Sciences
KIT103-0005
10014-582EA 418 USD InStock
10014-582
Adipogenesis Assay Kit, Enzo Life Sciences
Assays Cellular Assays
Convenient method for the enhanced induction and detection of adipogenesis, in the classic 3T3L-1 model.
For the induction and colorimetric detection of adipogenesis.
The Adipogenesis Assay Kit provides all the reagents needed to induce and detect adipogenesis. Optimized Differentiation, Insulin, and Enhancer solutions ensure maximal adipocyte induction. The Adipogenesis Dye allows for visual confirmation of differentiation and can also be easily extracted allowing measurement using a spectrophotometer or microplate reader.
Ordering information: Contains 1000x Differentiation Solution (Dexamethasone and IBMX), 1000x Insulin Solution, 1000x Enhancer Solution (Rosiglitazone), 1x Cell Fixative, 1x Adipogenesis Dye (Oil Red O), 1x Extraction Solution.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty