Would you like to log in again?
AMPIGENE® cDNA Synthesis Kit, Enzo Life Sciences
Supplier: Enzo Life Sciences
KIT106-0050 KIT106-0200
10014-592EA 1133.25 USD InStock
10014-592 10014-590
AMPIGENE® cDNA Synthesis Kit, Enzo Life Sciences
Nucleic Acid Reagents RT-PCR and cDNA Reagents
Efficient and sensitive cDNA synthesis, for downstream qPCR.
AMPIGENE® cDNA Synthesis Kit uses the latest developments in reverse transcriptase (RTase) technology and buffer chemistry for efficient and accurate cDNA synthesis.The modified MMLV RTase is blended with RNase inhibitor to prevent degradation of RNA by contaminating RNase. The RTase is thermostable and extremely active.An optimized buffer system contains oligo(dT), random hexamers, enhancers, dNTPs and MgCl2, allowing efficient and sensitive cDNA synthesis.
Ordering information: Contains 5x cDNA synthesis mix (anchored oligo (dT), random hexamers, 15mM MgCl2, 5mM dNTPs, enhancers, and stabilizers) 20x RTase (with RNase inhibitor).
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty