Would you like to log in again?
AMPIGENE® dNTP Mix, Enzo Life Sciences
Supplier: Enzo Life Sciences
AMPIGENE®
NUC102-0500
10014-628EA 67.71 USD InStock
10014-628
AMPIGENE® dNTP Mix, Enzo Life Sciences
Nucleic Acid Reagents Nucleotides
Ultra-pure, stable, and suitable for a variety of PCR applications.
AMPIGENE® dNTP Mix is an ultrapure mix of deoxyribonucleotide triphosphates to use as building blocks for a wide variety of PCR applications. It can withstand multiple freeze-thaw cycles and shown to retain 95% dNTPs in triphosphate form after 5 weeks at room temperature. AMPIGENE® dNTP Mix has been tested on many molecular biology applications including standard PCR, fast PCR, 1-step PCR, high-fidelity PCR, real-time PCR, lon-range PCR, MDA, RDA, LAMP, and DNA sequencing.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty